Learn More
Thermo Scientific™ Ropinirole hydrochloride
Chemical Identifiers
| CAS | 91374-20-8 |
|---|---|
| Chemical Name or Material | Ropinirole hydrochloride |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC465000010
|
Thermo Scientific™
465000010 |
1 g |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
AC465002500
|
Thermo Scientific™
465002500 |
250 mg |
N/A
|
N/A | |||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 91374-20-8 | |
| Ropinirole hydrochloride |
Specifications
| 91374-20-8 | |
| 1 g | |
| 241.0°C to 243.0°C | |
| Ropinirole hydrochloride |
Safety and Handling
GHS H Statement Harmful if swallowed.
Very toxic to aquatic life.
GHS P Statement IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face, hands and any exposed skin thoroughly after handling.
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
RUO â Research Use Only