Learn More
Thermo Scientific™ Rollout Drawers
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire refrigerator shelving.
Supplier:  Thermo Scientific™ 6704
 View more versions of this product
Catalog No. 361017657
Only null left
Description
Description
Choose stainless-steel Thermo Scientific™ Rollout Drawers as a convenient alternative to open-wire shelving. This is an option for Thermo Scientific™ high-performance refrigerators.
  • Stainless-steel drawers
  • Adjustable with full extension
Ordering Information:
Factory installed; must be specified when ordering the refrigerator
Specifications
Specifications
Complete SS Drawer Set
826 L
6
29.2 cu. ft.
Stainless Steel (Drawer)
29.2 cu. ft. (826L) Refrigerators and Freezers
Product Suggestions
Product Suggestions