missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Rolling Friction Carts - Set of 4

Catalog No. S000867
Change view
Click to view available options
Product Type:
Friction Cart
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S000867 Friction Cart
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65
Catalog No. S000867 Supplier United Scientific™ Supplier No. ROLLFRICT
Prop 65

This set of Rolling Friction Carts includes four carts of equal mass, each with wheels made from a different material.

This set includes four carts of equal mass, each with wheels made from a different material: soft silicone, soft rubber, wood, and fabric. The free rolling wheels quantitatively demonstrate friction without slipping.

  • Cart Dimensions: 5.25 x 2.125 x 1 in.
  • Well dimensions: 2.3 x 2 x 0.94 in.
  • Approximate wheel dimensions: 1.9 in. long, 1.17 in. diameter
  • Smooth rolling wheels
  • Four different materials
  • Includes activity guide

Specifications

Product Type Friction Cart
Dimensions (L x W x H) 5.25 x 2.125 x 1 in.