Learn More
Celltreat Scientific Products 490cm Tissue Culture Treated Roller Bottle, Non-Vented Cap, Sterile
Catalog No. 50201989
Encompass
CELLTREAT Scientific Products 490cm Roller Bottle. Large knurled cap for easy grip. Manufactured from medical grade virgin polystyrene. Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment. Easy-to-read black printed graduations. Non-vented cap. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile in individual bags, 24/case.
Catalog No. 50201989 Supplier Celltreat Scientific Products Supplier No. 229382
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CELLTREAT Scientific Products 490cm Roller Bottle. Large knurled cap for easy grip. Manufactured from medical grade virgin polystyrene. Tissue Culture Treated Roller Bottles have uniform hydrophilic surface ideal for consistent cell attachment. Easy-to-read black printed graduations. Non-vented cap. RNase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile in individual bags, 24/case.