Learn More
Brady™ Roll Form Pipe Markers, Legend: COMPRESSED AIR
Catalog No. 19111966
Change view
Click to view available options
Color:
Black, Yellow
White, Blue
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19111966 White, Blue
19111965 Black, Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19111966 Supplier Brady™ Supplier No. 41456
Ideal for identifying pipe content and flow without the hassle of precise measurement around the pipe's diameter. Brady Roll Form Pipe Marker, Legend: COMPRESSED AIR, is made of pressure sensitive vinyl.
  • Wrap around entire width of pipe and adhere to themselves
  • Legend repeats 65 times per roll
  • For marking pipes that carry same contents
  • With vinyl adhesive suited for use in outdoor applications
Specifications
Color White, Blue
Material Pressure-sensitive Vinyl
Product Type Roll Form Pipe Marker
Display Text Legend: COMPRESSED AIR
Quantity 1 Roll
Size (English) 30 ft.x 8 in.
Size (Metric) 9.14 m x 20.3 cm