Learn More
Brady™ Roll Form Pipe Marker: CHILLED WATER
Catalog No. 19112286
Change view
Click to view available options
Display Text:
CHILLED WATER
Catalog No. Display Text
19112286 CHILLED WATER
1 options
Catalog No. 19112286 Supplier Brady™ Supplier No. 41518
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Provide clear notice to workers by identifying pipe contents and flow without the hassle of precise measurement around a pipe's diameter. Brady™ Roll Form Pipe Markers are made of Pressure Sensitive Vinyl and display text with clear color contrast.
  • Made of self-sticking vinyl suitable for indoor or outdoor applications
  • Legend repeats 65 times per roll
Specifications
Adhesive Type Acrylic, Permanent, Cold-temperature
Color White, Green
Format Roll
Material Pressure-sensitive Vinyl (B-946)
Product Type Roll Form Pipe Marker
Display Text CHILLED WATER
Size (English) 30 ft. x 8 in.
Thickness (English) 0.004 in.