missing translation for 'onlineSavingsMsg'
Learn More

BD RODAC™ SNAP LID Trypticase™ Soy Agar (TSA) w Lecithin, Polysorbate 80 (LP80), Penicillinase

Catalog No. B215274
Change view
Click to view available options
Quantity:
100/Pk.
Packaging Type:
Sterile Double Wrapping Plate
Triple Wrapping Isolator Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
B215274 100/Pk. Sterile Double Wrapping Plate
B215273 100/Pk. Triple Wrapping Isolator Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B215274 Supplier BD Supplier No. 215274

The BD RODAC SNAP LID is a RODAC contact plate equipped with a unique SNAP LID locking mechanism to provide additional security during sampling and transport from a controlled environment to the laboratory.

The RODAC SNAP LID has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate; that:

  • Withstands an accidental drop from the height of a bench-top workspace
  • Ensures the integrity of the sample
  • Prevents the dish from opening unintentionally
  • Protects media from accidental cross contamination
  • Improves stability during stacking and storage

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Light Beige
Form Liquid
Manufacturing Origin United States
Media Type Trypticase™ Soy Agar with Lecithin, Polysorbate 80 and Penicillinase
Sterility Sterile
For Use With (Application) Rodac™ snap lid has a latch on the base of the plate that engages the ridge on the lid of the plate to lock the plate
Packaging Type Sterile Double Wrapping Plate
Quantity 100/Pk.