missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Rocker Cell Culture Bags

Catalog No. MT9120092
Change view
Click to view available options
Capacity (Metric):
2 L
10 L
20 L
22 L
50 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
MT9120092 22 L
MT9120079 10 L
MT9120081 50 L
MT9120080 2 L
MT9120078 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MT9120092 Supplier Corning™ Supplier No. 9120092

Features a multilayered film with an inert PE (Polyethylene) fluid contact layer

Our Rocker Cell Culture Bag, irradiated, features ethylene vinyl acetate/low-density polyethylene 9101 film for use with commercial rocking-motion bioreactor chambers. The 9101 film is clear, multi-layered single web film with a co-polymer contact surface. Corning offers this single-use cell culture bag in 2L-50L configurations. The Rocker Cell Culture Bag features bag ports with a 1/4” barb connected to a pall filter and check valve with small slide clamps. The bag ports with a 1/8” barb are connected to the male MPC plug and male Luer plug with female MPC and female Luer arrangements. The male Luer plug (without a stem) and swabable valve are connected to the remaining bag port with a 1/8” barb. The rocking-motion of the bioreactor platform creates effective, non-invasive mixing of the cell culture/media with efficient gas transfer. Rocking-motion prevents cells from settling at the bottom of the Rocker Cell Culture Bag while generating large amounts of recombinant protein in both mammalian and insect cell lines. The closed-system eliminates the risk of cross-contamination. The stability bars located on each side allow commercially available bioreactors to accommodate the cell culture bag. This single-use cell culture bag is ideal for basic research to large-scale biopharmaceutical manufacturing applications.
  • Complete Certificate of Compliance is available for each production lot.
  • P1 Check valve with 1/4' barb PVDF 8004022 filter 2' length 3/16' I.D. x 3/8' O.D. PCMGS tube
  • P2 PVDF 8004022 filter 2' length 3/16' I.D. x 3/8' O.D. PCMGS tube
  • P3 Female MPC connector with 1/4' barb Male MPC plug 40' length 1/4' I.D. x 7/16' O.D. chemically resistant, heat sealable flexible tube
  • P4 Female Luer with 1/4' barb Male Luer plug 40' length 1/8' I.D. x 1/4' O.D. chemically resistant, heat sealable flexible tube
  • P5 Female Luer with 1/4' barb Swabbable valve Male Luer plug without stem
  • Irradiated EVA/LDPE Film (9101)
Corning media certificates are available on our Resource Library.

Specifications

Content And Storage 10° to 34°C
Certifications/Compliance ANSI, AMI, ISO 11137
For Use With (Application) Commercial rocking-motion bioreactor chambers by providing non-invasive mixing of the cell culture/media with efficient gas transfer and ideal for basic research to large-scale biopharmaceutical manufacturing applications
Tubing Style Sealable flexible tubing
Film Description EVA/LDPE 9101 film is clear, multi-layered single web film with a copolymer contact surface
No. per Case 1/Ea.
No. per Pack 1/Pk.
Port Detail Port 1 - 0.25 in. barb connected to a pall filter and check valve with small slide clamps, 2 L x 0.18 I.D. x 0.37 in. O.D. PC mgS tubing, port 2 - 0.25 in. Barb connected to a pall filter with small slide clamps, 2 L x 0.18 I.D. x 0.37 in. O.D. PC mgS tubin
Quantity Each
Sterilization Method Radiation
Product Type Cell Culture Bag
Capacity (Metric) 22 L
Show More Show Less