Learn More
Rock Formation Charts
Catalog No. S45018A
Change view
Click to view available options
Product Type:
Igneous Rock Chart
Metamorphic Rock Reference Sheet
Metamorphic Rocks Chart
Sedimentary Rock Reference Sheet
Sedimentary Rocks Chart
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
S45018A Igneous Rock Chart
S45002 Sedimentary Rocks Chart
S45002A Sedimentary Rock Reference Sheet
S45004A Metamorphic Rock Reference Sheet
S45004 Metamorphic Rocks Chart
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S45018A Supplier Geoscience Industries Supplier No. S45018A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This coordinated series of display charts describes the classification, identification, and formation of the three major rock types. each chart also includes representative specimens of the appropriate rock group. For each of the charts, sets of 15 notebook-sized student reference sheets are also available.
Igneous Rocks Chart
A systematic arrangement of igneous rocks according to texture, composition, and origin. Includes 33 rock specimens.
Metamorphic Rocks Chart
The intricate processes by which a parent rock or mineral is changed (metamorphosed) to a new form are graphically illustrated. Includes 26 rock and mineral specimens.
Sedimentary Rocks Chart
The three main categories of sedimentary rocks (clastic, chemical, and organic) are clearly demonstrated with emphasis on the importance of origin and particle size. Includes 21 rock specimens.
Order Info
Measuring 60 x 39cm and mounted in wood frames.
Specifications
Product Type Igneous Rock Chart
Includes 26 Rock and Mineral specimens, notebook reference sheets
Subject Science