Learn More
Fluotics Agilent Bravo compatible P70, sterile
Catalog No. NC1924450
Encompass
Agilent Bravo compatible P70, sterile, working volume: 70 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 19133-212
Catalog No. NC1924450 Supplier Fluotics Supplier No. AGI70.ST
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Agilent Bravo compatible P70, sterile, working volume: 70 uL, Case Size: 50 racks/case with 384 tips per rack, Compare to: 19133-212