Learn More
Stemcell Technologies RoboSep™ PE Positive Selection Kit II, For processing: 1 x 10^9 cells DFS Item
Supplier:  Stemcell Technologies Inc 17684RF
The EasySep™ PE Positive Selection Kit II is designed to isolate cells that are labeled with PE-conjugated antibodies. Desired cells are targeted with antibody complexes recognizing PE and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off.This product replaces the EasySep™ PE Positive Selection Kit (Catalog #18557) for even faster cell isolations.
Catalog No. 501974554
Only null left
Explore available promotions