missing translation for 'onlineSavingsMsg'
Learn More

Colloidal Silicon Dioxide, NF, EP, 99-100.5%, Spectrum™ Chemical

Catalog No. 18601818
Change view
Click to view available options
Quantity:
100 g
500 g
5 kg
10 kg
Packaging:
Fiber Drum
Plastic Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18601818 100 g Plastic Bottle
18601819 500 g Poly Pail
18601821 5 kg Fiber Drum
18601820 10 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18601818 Supplier Spectrum Chemical Mfg Cor Supplier No. S1510100GM
Get Up to $300 in Additional Chemicals
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

S1510, 7631-86-9, SiO2

Spectrum™ Chemical Colloidal Silicon Dioxide, NF, EP is used both in the food and pharmaceutical industries as a powder flow helper in the production of tablets. All Spectrum Chemical NF, EP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7631-86-9
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

CAS Max % 100%
Material Silicon Dioxide
Quantity 100 g
Loss on Drying 2.5%
Loss on Ignition 5%
Packaging Plastic Bottle
Percent Purity 99 to 100.5%
Grade Multi-Compendial
Chemical Name or Material Colloidal Silicon Dioxide