missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ RNA Polymerases

Catalog No. FEREP168AF
Change view
Click to view available options
Polymerase:
Poly(A) RNA Polymerase
T7 RNA Polymerase
Concentration:
200 U/μL
5 U/μL
Quantity:
20,000 U
800 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Polymerase Concentration Quantity
FEREP168AF Poly(A) RNA Polymerase 5 U/μL 800 U
FEREP0111AF T7 RNA Polymerase 200 U/μL 20,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FEREP168AF Supplier Thermo Scientific™ Supplier No. EP168AF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Thermo Scientific enzymes for in vitro transcription are designed for efficient and reliable RNA synthesis. These enzymes cover the entire synthesis workflow from template preparation to post-transcriptional modifications, ensuring optimal performance and high yields of synthesized RNA.

High quality RNA synthesized by in vitro transcription is essential for RNA research and this versatile technique can be used in a variety of applications including the introduction of modified nucleotides into RNA transcripts for enhanced translational capacity and reduced immune activity of the cells.

Features

  • Convenient stand-alone enzymes and buffers for workflow optimization
  • No animal-origin components in final product composition
  • Reliable and reproducible results with rigorous quality testing
  • Scalable and available as OEM products

Applications

  • CRISPR/Cas-based genome editing
  • Gene expression
  • Generation of pluripotent cells

Specifications

Content And Storage 5 U/μL Poly(A) Polymerase, 160 μL

Store at -20°C.
Polymerase Poly(A) RNA Polymerase
Product Line TheraPure
For Use With (Application) RNA Polyadenylation
Quantity 800 U
Concentration 5 U/μL

For Research Use Only. Not for use in diagnostic procedures.