Learn More
Medicom™ Ritmed™ Laparotomy Sponges, Tape Loop, Blue Crimped Paper
Catalog No. 17377448
Change view
Click to view available options
Style:
Non-washed, Tape Loop
Pre-washed, Tape Loop
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Style
17377448 Pre-washed, Tape Loop
17377449 Non-washed, Tape Loop
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17377448 Supplier Medicom™ Supplier No. MED1525
Only null left
Laparotomy sponges are surgical sponges used to absorb fluids, maintain a clear surgical field, and protect tissues during procedures such as abdominal and thoracic surgeries
  • Ritmed™ Laparotomy sponges are made of 100% cotton gauze.
  • 4-Ply with high absorbency and soft texture.
  • X-ray Detectable for use in the operation room.
  • Not made with Natural Rubber Latex.
Specifications
Material Cotton
Product Type Surgical Gauze Sponge
Sterility Sterile
Size Universal
Dimensions (L x W) 4 x 18 in.
Latex Free Yes
Style Pre-washed, Tape Loop
Autoclavable Not Autoclavable