missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ritalinic Acid 98.0+%, TCI America™
$154.71 - $505.16
Chemical Identifiers
| CAS | 19395-41-6 |
|---|---|
| Molecular Formula | C13H17NO2 |
| Molecular Weight (g/mol) | 219.284 |
| MDL Number | MFCD06200695 |
| InChI Key | INGSNVSERUZOAK-UHFFFAOYSA-N |
| Synonym | 2-Phenyl-2-(2-piperidyl)acetic Acid |
| PubChem CID | 86863 |
| ChEBI | CHEBI:83481 |
| IUPAC Name | 2-phenyl-2-piperidin-2-ylacetic acid |
| SMILES | C1CCNC(C1)C(C2=CC=CC=C2)C(=O)O |
Chemical Identifiers
| 19395-41-6 | |
| 219.284 | |
| INGSNVSERUZOAK-UHFFFAOYSA-N | |
| 86863 | |
| 2-phenyl-2-piperidin-2-ylacetic acid |
| C13H17NO2 | |
| MFCD06200695 | |
| 2-Phenyl-2-(2-piperidyl)acetic Acid | |
| CHEBI:83481 | |
| C1CCNC(C1)C(C2=CC=CC=C2)C(=O)O |
Specifications
| 19395-41-6 | |
| White-Yellow | |
| MFCD06200695 | |
| 2-Phenyl-2-(2-piperidyl)acetic Acid | |
| C1CCNC(C1)C(C2=CC=CC=C2)C(=O)O | |
| 219.284 | |
| CHEBI:83481 | |
| ≥98.0% (HPLC,T) | |
| Ritalinic Acid |
| 245°C | |
| C13H17NO2 | |
| 1 g | |
| INGSNVSERUZOAK-UHFFFAOYSA-N | |
| 2-phenyl-2-piperidin-2-ylacetic acid | |
| 86863 | |
| 219.28 | |
| Crystalline Powder |
Safety and Handling
TSCA : No