missing translation for 'onlineSavingsMsg'
Learn More

Ritalinic Acid 98.0+%, TCI America™

Catalog No. R01111G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01111G 1 g
R01115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01111G Supplier TCI America Supplier No. R01111G

Chemical Identifiers

CAS 19395-41-6
Molecular Formula C13H17NO2
Molecular Weight (g/mol) 219.284
MDL Number MFCD06200695
InChI Key INGSNVSERUZOAK-UHFFFAOYSA-N
Synonym 2-Phenyl-2-(2-piperidyl)acetic Acid
PubChem CID 86863
ChEBI CHEBI:83481
IUPAC Name 2-phenyl-2-piperidin-2-ylacetic acid
SMILES C1CCNC(C1)C(C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 245°C
Color White-Yellow
Quantity 1 g
Formula Weight 219.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Ritalinic Acid