missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals RITA, p53 activator III
A cell-permeable, p53-targeting, tricyclic thiophene derivative
$534.48
Chemical Identifiers
| CAS | 213261-59-7 |
|---|---|
| Molecular Formula | C14H12O3S2 |
| Synonym | 5,5′-(2,5-Furandiyl)bis-2-thiophenemethanol; NSC 652287 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AAJ65379MA
|
Thermo Scientific Chemicals
J65379MA |
10 mg |
Each for $534.48
|
|
|||||
Description
A cell-permeable, p53-targeting, tricyclic thiophene derivative
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 213261-59-7 | |
| C14H12O3S2 | |
| 5,5′-(2,5-Furandiyl)bis-2-thiophenemethanol; NSC 652287 |
Specifications
| 2°C to 8°C | |
| 213261-59-7 | |
| 292.40g/mol | |
| Light Sensitive | |
| Crystalline solid | |
| OCC1=CC=C(S1)C1=CC=C(O1)C1=CC=C(CO)S1 | |
| 292.37 |
| A cell-permeable, p53-targeting, tricyclic thiophene derivative | |
| C14H12O3S2 | |
| 5,5′-(2,5-Furandiyl)bis-2-thiophenemethanol; NSC 652287 | |
| Signal Transduction Reagent | |
| KZENBFUSKMWCJF-UHFFFAOYSA-N | |
| (5-{5-[5-(hydroxymethyl)thiophen-2-yl]furan-2-yl}thiophen-2-yl)methanol |
Safety and Handling
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only