missing translation for 'onlineSavingsMsg'
Learn More

Risperidone, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 459390050

 View more versions of this product

Catalog No. AC459390050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Risperidone
0.5% max.
98.5% min. (HPLC)
5g
RAPZEAPATHNIPO-UHFFFAOYSA-N
3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
5073
410.49
106266-06-2
Authentic
C23H27FN4O2
risperidone, risperidal, risperdal, risperdal consta, rispolept, risperin, rispolin, sequinan, risperidona, risperidonum
CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
410.49
CHEBI:8871
99%

Chemical Identifiers

106266-06-2
410.49
risperidone, risperidal, risperdal, risperdal consta, rispolept, risperin, rispolin, sequinan, risperidona, risperidonum
CHEBI:8871
CC1=C(C(=O)N2CCCCC2=N1)CCN3CCC(CC3)C4=NOC5=C4C=CC(=C5)F
C23H27FN4O2
RAPZEAPATHNIPO-UHFFFAOYSA-N
5073
3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.

GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only