Learn More
Ansell RINGERS™ R167 TPR-designed Impact Gloves
Touchscreen-compatible impact and cut protection gloves, for hand safety and ease of use.
Supplier: Ansell 16711
Description
- Certified ANSI/ISEA 138 Level 1 Flexible knuckle TPR design with two detached fingers Durable synthetic
- Developed with Ringers' F3 Technology™ and patented TPR design for superior dexterity and comfort
- Touchscreen compatible index, middle and thumb tips lessen the need to remove gloves
- High visibility and hook and loop pull tab closure provides added security on the job
Specifications
| Leather | |
| ANSI A3 | |
| 11 |
| Yellow | |
| Yes | |
| Ringers™ R-167 |