missing translation for 'onlineSavingsMsg'
Learn More

Ansell RINGERS™ R075 Red Dipped Gloves

Catalog No. p-200008705
Change view
Click to view available options
Size:
10
12
8
Catalog No. Size
17000657 8
17000658 10
17000659 12
3 options

Catalog No. 17000657

Supplier: Ansell 07508

Only null left

Waterproof PVC-coated long gauntlet-style impact gloves, for advanced chemical protection

  • Advanced defenses: RINGERS™ R075 impact protection gloves' TPR (thermoplastic rubber) design, developed with F3 Technology™, comfortably protects the top of the hand and full length of the fingers
  • Specialized protection: Tested against EN 374-1 Type A chemical resistance standards, these RINGERS™ gloves guard against various hazards*
  • Added safety: With a protective inner liner, they satisfy EN 388 level C and ANSI/ISEA level A3 cut resistance standards
  • Enhanced grip and durability: Grip and durability are also enhanced by these chemical-resistant gloves’ textured palm and PVC coating, resulting in EN 388 level 4 and ANSI/ISEA level 5 abrasion resistance
  • Certified heat resistance: These impact gloves also provide EN 407 level 1 contact heat resistance**

*CE-rated for EN 374-1: A (Methanol) J (n-Heptane) K (Sodium Hydroxide 40%) L (Sulfuric Acid 96%) M (Nitric Acid 65%) O (Ammonium Hydroxide 25%)

**Withstanding outside exposure up to 100°C/212°F

  • RECOMMENDED FOR
    • Drilling assembly and completion
    • Handling jagged metal
    • Mixing or loading chemicals
    • Body painting including surface inspection
    • Applying sealers, touching up
    • Oil, fluids and filter change
    • Spill or leakage cleanup
    • Plating, coating, sealing, painting
    • Hydraulic repair
    • Use of binding agents
    • Smelting and cleaning of equipment
    • Re-fuel oil fluid and diesel
    • Filling, blending and charging of raw materials
    • Opening furnaces draining pumps valves or lines and crackers BTX process
    • Unexpected leakages spills or other releases

Specifications

ANSI Abrasion Rating ANSI 2
Chlorinated No
Applications Industrial
Color Red
Latex Free No
Material PVC
Size 8
Glove Cuff Style Straight
Lining Lined