missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ringer's Solution, lactate-buffered

Catalog No. AAJ67572AP
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67572AP 500 mL
AAJ67572K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67572AP Supplier Thermo Scientific Chemicals Supplier No. J67572AP
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Name or Material Ringer's Solution
Physical Form Liquid
Quantity 500 mL
Synonym dihydrogen oxide, dihydrogen monoxide
Packaging Plastic bottle

RUO – Research Use Only