Learn More
American Scientific Ring and Disc Moment of Inertia Apparatus
Use this item for studying the dependence of the moment of inertia on the distribution of mass
Supplier: American Scientific 347900
Description
- Roll a metal ring and wooden disk of the same mass and diameter on an inclined plane or a level surface; the moment of inertia is displayed!
- The product is comprised of a metal ring and a wooden disk of diameter 6 in. (150 mm), 2 in. (50 mm) thick and both the same mass
- When these are rolled down on an inclined plane or on a level surface with the same initial impulse, the difference in acceleration can be studied
Specifications
| Energy, Motion, Inertia | |
| Metal ring and wooden disc |