missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ 96-Well Rigid Frame PCR Plates, BIO-CERT™ PCR QUALITY

For medium and high sample throughput, these 96-well plates are an efficient solution for PCR and qPCR.

Supplier:  BRANDTECH™ BRAND™ BRT781542

 View more versions of this product

Catalog No. 01213142


Only null left

Description

Description

BRAND PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:

    Exceptional Quality and Purity
  • BIO-CERT™ PCR QUALITY guarantees plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
  • Manufactured without contaminants that could interfere with reagents or samples
  • Ultra-smooth vessel interiors minimize enzyme and nucleic acid binding for reliable results
    Robust Design and Durability
  • Strong polycarbonate frames provide safe handling in both semi-automated and automated pipetting systems
  • Ensures optimal evaporation protection during thermal cycling
    Precision Thermal Performance
  • Ultra-thin-walled wells made from 100% virgin medical-grade polypropylene
  • Uniform and thin wall thickness promotes rapid and consistent heat transfer, reducing cycle times
    Versatile Compatibility
  • Available with multiple skirt options for a secure fit in most common thermal cyclers
    Optimized for Workflow Efficiency
  • Plates are available in various colors to help distinguish workflow steps
  • Alphanumeric coding ensures easy and accurate sample identification
  • White plates are specifically designed for qPCR, providing optimal fluorescence signal reflection
Specifications

Chemical Identifiers

No Barcode
BIO-CERT PCR Quality
8 x 12
Bagged
Polycarbonate, Polypropylene
Non-sterile
Conical Bottom (V-well)
PCR Ready
Untreated
Standard
Autoclavable
Single-use
96
Non-treated
26 cm

Specifications

No Barcode
BIO-CERT PCR Quality
PCR plate 96-well, Rigid Frame, PC/PP, golden, full skirted, Low Profile, wells transparent, BIO-CERT™ PCR QUALITY
Autoclavable
Gold
PCR 96-well plate
Full
0.15 mL
Non-sterile
Conical Bottom (V-well)
Transparent
26 cm
PCR Ready
Untreated
Standard
8 x 12
Bagged
Single-use
PCR Cycler
Polycarbonate, Polypropylene
96
Non-treated
Low Profile
26 cm
13 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions