missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ BRAND™ 96-Well Rigid Frame PCR Plates, BIO-CERT™ PCR QUALITY
For medium and high sample throughput, these 96-well plates are an efficient solution for PCR and qPCR.
Supplier: BRANDTECH™ BRAND™ 781548
Description
BRAND PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:
- Exceptional Quality and Purity
- BIO-CERT™ PCR QUALITY guarantees plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
- Manufactured without contaminants that could interfere with reagents or samples
- Ultra-smooth vessel interiors minimize enzyme and nucleic acid binding for reliable results
- Robust Design and Durability
- Strong polycarbonate frames provide safe handling in both semi-automated and automated pipetting systems
- Ensures optimal evaporation protection during thermal cycling
- Precision Thermal Performance
- Ultra-thin-walled wells made from 100% virgin medical-grade polypropylene
- Uniform and thin wall thickness promotes rapid and consistent heat transfer, reducing cycle times
- Versatile Compatibility
- Available with multiple skirt options for a secure fit in most common thermal cyclers
- Optimized for Workflow Efficiency
- Plates are available in various colors to help distinguish workflow steps
- Alphanumeric coding ensures easy and accurate sample identification
- White plates are specifically designed for qPCR, providing optimal fluorescence signal reflection
Chemical Identifiers
| No Barcode | |
| BIO-CERT PCR Quality | |
| 8 x 12 | |
| Bagged | |
| Polycarbonate, Polypropylene | |
| Non-sterile | |
| Conical Bottom (V-well) | |
| PCR Ready |
| Untreated | |
| Standard | |
| Autoclavable | |
| Single-use | |
| 96 | |
| Non-treated | |
| 26 cm |
Specifications
| No Barcode | |
| BIO-CERT PCR Quality | |
| PCR plate 96-well, Rigid Frame, PC/PP, blue, semi skirted, Standard, wells transparent, BIO-CERT™ PCR QUALITY | |
| Autoclavable | |
| Blue | |
| PCR 96-well plate | |
| Semi | |
| 0.2 mL | |
| Non-sterile | |
| Conical Bottom (V-well) | |
| Transparent | |
| 22 cm | |
| PCR Ready |
| Untreated | |
| Standard | |
| 8 x 12 | |
| Bagged | |
| Single-use | |
| PCR Cycler | |
| Polycarbonate, Polypropylene | |
| 96 | |
| Non-treated | |
| Standard | |
| 26 cm | |
| 10.5 cm |