missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ 96-Well Rigid Frame PCR Plates, BIO-CERT™ PCR QUALITY

Catalog No. 01213132
Change view
Click to view available options
Color:
Blue
Frosted
Gold
Green
Red
Transparent
White
Skirt Style:
Full
Semi
Profile:
Low Profile
Standard
Volume (Metric) Well:
0.15 mL
0.2 mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Skirt Style Profile Volume (Metric) Well
01213132 Blue Full Low Profile 0.15 mL
01213135 Green Full Low Profile 0.15 mL
01213134 White Full Low Profile 0.15 mL
01213137 Blue Semi Standard 0.2 mL
01213141 Frosted Full Low Profile 0.15 mL
01213142 Gold Full Low Profile 0.15 mL
01213136 Transparent Semi Standard 0.2 mL
01213139 Transparent Semi Low Profile 0.15 mL
01213133 Red Full Low Profile 0.15 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 01213132 Supplier BRANDTECH™ BRAND™ Supplier No. 781545
Only null left

For medium and high sample throughput, these 96-well plates are an efficient solution for PCR and qPCR.

BRAND PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:

    Exceptional Quality and Purity
  • BIO-CERT™ PCR QUALITY guarantees plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
  • Manufactured without contaminants that could interfere with reagents or samples
  • Ultra-smooth vessel interiors minimize enzyme and nucleic acid binding for reliable results
    Robust Design and Durability
  • Strong polycarbonate frames provide safe handling in both semi-automated and automated pipetting systems
  • Ensures optimal evaporation protection during thermal cycling
    Precision Thermal Performance
  • Ultra-thin-walled wells made from 100% virgin medical-grade polypropylene
  • Uniform and thin wall thickness promotes rapid and consistent heat transfer, reducing cycle times
    Versatile Compatibility
  • Available with multiple skirt options for a secure fit in most common thermal cyclers
    Optimized for Workflow Efficiency
  • Plates are available in various colors to help distinguish workflow steps
  • Alphanumeric coding ensures easy and accurate sample identification
  • White plates are specifically designed for qPCR, providing optimal fluorescence signal reflection

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance BIO-CERT PCR Quality
Format Standard
Description PCR plate 96-well, Rigid Frame, PC/PP, blue, full skirted, Low Profile, wells transparent, BIO-CERT™ PCR QUALITY
Array 8 x 12
Autoclavable Autoclavable
Packaging Bagged
Color Blue
Disposable Single-use
Product Type PCR 96-well plate
For Use With (Equipment) PCR Cycler
Skirt Style Full
Material Polycarbonate, Polypropylene
Volume (Metric) Well 0.15 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Profile Low Profile
Well Color Transparent
Length (Metric) 26 cm
Width (Metric) 26 cm
Height (Metric) 13 cm
Fit for Purpose PCR Ready
Show More Show Less