missing translation for 'onlineSavingsMsg'
Learn More

Rifaximin 97.0+%, TCI America™

Catalog No. p-7135695
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01011G 1 g
R01015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R01011G Supplier TCI America Supplier No. R01011G

Chemical Identifiers

CAS 80621-81-4
Molecular Formula C43H51N3O11
Molecular Weight (g/mol) 785.89
MDL Number MFCD00864973
InChI Key NZCRJKRKKOLAOJ-XRCRFVBUSA-N
Synonym rifaximin
PubChem CID 72698618
IUPAC Name (7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z)-2,15,17,36-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22,30-octamethyl-6,23-dioxo-8,37-dioxa-24,27,33-triazahexacyclo[23.10.1.1⁴,⁷.0⁵,³⁵.0²⁶,³⁴.0²⁷,³²]heptatriaconta-1,3,5(35),9,19,21,25(36),26(34),28,30,32-undecaen-13-yl acetate
SMILES CO[C@H]1\C=C\O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C1=C(N=C5C=C(C)C=CN15)C4=C3C2=O

Specifications

Melting Point 212°C
Quantity 1 g
Formula Weight 785.89
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Rifaximin
RTECSNumber KD1576000
TSCA No
Recommended Storage Refrigerator