missing translation for 'onlineSavingsMsg'
Learn More

Rifamycin SV sodium, 98+%

Catalog No. AAJ6309503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6309503 1 g
AAJ6309506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6309503 Supplier Thermo Scientific Chemicals Supplier No. J6309503

CAS: 14897-39-3 | C37H47NNaO12 | 720.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Solubility
Soluble in water, alcohol and dimethyl sulfoxide.

Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 14897-39-3
Molecular Formula C37H47NNaO12
Molecular Weight (g/mol) 720.77
MDL Number MFCD09752825
InChI Key YVOFSHPIJOYKSH-DITFEXNXSA-N
PubChem CID 124219641
IUPAC Name sodium (7S,9Z,11S,12R,13S,14R,15R,16R,17S,18S,19Z,21Z)-2,15,17,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29),26-octaen-13-yl acetate
SMILES [Na+].CO[C@H]1\C=C/O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C\[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)C=C(O)C4=C3C2=O

Specifications

Type Rifamycin SV Sodium
Physical Form Powder
Color Orange to Red
Quantity 1 g
Merck Index 14,8218
Solubility Information Soluble in water,alcohol and dimethyl sulfoxide.
Formula Weight 719.45
Percent Purity ≥98%
Chemical Name or Material Rifamycin SV sodium
EINECSNumber 238-965-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only