missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rifampin, Molecular Biology Reagent

Catalog No. AAJ6083603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6083603 1 g
AAJ6083606 5 g
AAJ6083614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6083603 Supplier Thermo Scientific Chemicals Supplier No. J6083603

Rifampin (rifampicin), CAS # 13292-46-1, is a synthetic rifamycin antibiotic that works by inhibiting RNA and DNA synthesis and exhibits activity against the tuberculosis microbe. | CAS: 13292-46-1 | C43H58N4O12 | 822.953 g/mol

Antibiotic effective against mycobacteria. Can also inhibit DNA polymeraseRifampin is an antibiotic used in the treatment of tuberculosis and its related microbacterial infections. It is involved in the inhibition of DNA polymerase and RNA polymerase. As an antipruritic agent, it is used in the autoimmune cholestatic liver disease and primary biliary cirrhosis (PBC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Rifampin is a rifamycin class antibiotic synthesized to resemble the compound isolated from Amycolatopsis rifamycinica

Applications

  • Rifampin can be used as a bacteriostatic that prevents RNA and DNA synthesis, particularly in mycobacteria
  • It may also act as an antineoplastic, a neuroprotective, and inhibitor of angiogenesis

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
MDL Number MFCD00151389
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym Rifampicin
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Type Rifampicin
Physical Form Powder
Melting Point 183°C to 188°C
Color Brown/Red to Orange
Quantity 1 g
Beilstein 5723476
Merck Index 14,8216
Solubility Information Soluble in DMSO or methanolSoluble in water,ethyl acetate,chloroform,methanol,tetrahydrofuran and dimethyl sulfoxide.
Formula Weight 822.94
Grade Molecular Biology
Chemical Name or Material Rifampin
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 236-312-0
RTECSNumber VJ7000000
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only