missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Rifampicin, Calbiochem™,

Catalog No. 5573031GM
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
5573031GM 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 5573031GM Supplier MilliporeSigma™ Supplier No. 5573031GM

Recommended Storage

15°C to 30°C, DMSO stock solutions are Stable for up to 2 months at −20°C

Chemical Identifiers

CAS 13292-46-1
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.953
InChI Key FZYOVNIOYYPUPY-HRJPTAQKSA-N
Synonym rifampicin
PubChem CID 131839595
SMILES CC1C=CC=C(C(=O)NC2=C(C3=C(C(=C4C(=C3C(=O)C2=CNN5CCN(CC5)C)C(=O)C(O4)(OC=CC(C(C(C(C(C(C1O)C)O)C)OC(=O)C)C)OC)C)C)O)O)C

Specifications

Color Red
Quantity 1 g
Physical Form Solid
Recommended Storage 15°C to 30°C, DMSO stock solutions are Stable for up to 2 months at −20°C