missing translation for 'onlineSavingsMsg'
Learn More

Pyramex™ Ridgeline™ Full Brim Hard Hat

Catalog No. 19155464
Change view
Click to view available options
Color:
Matte Black Graphite
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19155464 White
19155468 Matte Black Graphite
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19155464 Supplier Pyramex™ Supplier No. HP54110

Ridgeline™ Full Brim Hard Hat is strong, yet ultra-light for seemingly weightless protection.

  • Shell constructed from ABS materials
  • Ratchet suspension is easy to adjust and allows the wearer to modify the
    fit while wearing hard hat
  • Replaceable suspensions and headbands available
  • 4-point suspension may be converted to 6-point suspension with
    optional strap (HP6PTSUS)
  • Custom imprinting available on hard hats

Specifications

Certifications/Compliance ANSI Z89.1 standards, Type 1 --- Class C, G, and E,CSA Z94.1-05
Color White
For Use With (Application) Construction
Material Shell: ABS, Suspension: HDPE and high impact Nylon, Sweat band: Polyester Fabric and Polyurethane Foam
Product Type 4-Point Ratchet
Size 6.5 to 8
Weight (Metric) 378 g
Ventilated Not Vented
Head Style Full Brim