Learn More
R-Biopharm Inc RIDA QUICK GLIADIN
Supplier: R-Biopharm GmbH R7003
RIDA QUICK Gliadin can be used as a swab test for the gluten determination on surfaces in the hygiene control and for the qualitative detection of gliadin/gluten in raw material. The test should only be used for the detection of small amounts of gluten (contaminations). No hook-effect is observed at high concentrations. The gliadin/gluten determination can be carried out for numerous raw materials after a simple ethanol extraction. The monoclonal antibody R5 reacts with the gliadinfraction from wheat and corresponding prolamins from rye and barley. 25 x dip sticks (one for each determination).