Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
Catalog No. 361014309
Change view
Click to view available options
Product Type:
Epredia Three Step Stain Kit
Solution A
Solution B
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
361014309 Epredia Three Step Stain Kit
22050275 Solution B
22050274 Solution A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 361014309 Supplier Epredia™ Supplier No. SH3300
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
  • Stain can be used to detect H. Pylori.
  • Set contains easy-to-follow instructions
  • Product is available in one-gallon containers to facilitate use on automated strainers
  • Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)
Specifications
Product Type Epredia Three Step Stain Kit