Learn More
Epredia™ Richard-Allan Scientific™ Three-Step Stain Kit
Obtain results on par with Wright-Giemsa with Epredia™ Richard-Allan Scientific™ Three Step Stain.
Supplier: Epredia™ SH3313
Includes: A package insert is included with easy-to-follow instructions.
Description
- Stain can be used to detect H. Pylori.
- Set contains easy-to-follow instructions
- Product is available in one-gallon containers to facilitate use on automated strainers
- Set contains three 500mL bottles consisting of Fixative Solution, Solution A(Eosin-Y), and Solution B(Azure A and Methylene Blue)
Order Info
Components are also available in one-gallon containers to facilitate use on automated stainers.
Specifications
| 1 gal. | |
| Solution A | |
| Stain set provides results equal to wright-giemsa | |
| A package insert is included with easy-to-follow instructions |