Learn More
Epredia™ Neg-50™ Frozen Section Medium
Catalog No. 22050454
Change view
Click to view available options
Quantity:
4 oz. per bottle
4 oz. per bottle, 2/cs.
4 oz. per bottle, 6/cs.
4 oz.per bottle
Color:
Blue
Clear
Five Colors plus Clear
Green
Orange
Pink
Yellow
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Color
22050454 4 oz. per bottle Yellow
361014314 4 oz. per bottle, 2/cs. Clear
22050451 4 oz.per bottle Blue
361014361 4 oz. per bottle Green
22046512 4 oz. per bottle Orange
22050453 4 oz. per bottle Pink
22046511 4 oz. per bottle, 6/cs. Clear
22050450 4 oz. per bottle Five Colors plus Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 22050454 Supplier Epredia™ Supplier No. 6502Y
Only null left
Yield high-quality sections down to -50°C without chatter using water-soluble Epredia™ Neg-50™ Frozen Section Medium. Comes in clear and five additional colors with all leaving no residue after staining.
Yield high-quality sections down to -50°C without chatter using Epredia; Neg-50™ Frozen Section Medium, a water-soluble frozen section medium.
  • Buffered to ensure proper alkalinity (pH = 8.0); leaves no residue after staining
  • Specially formulated to prevent tissue separation from the slide
Ordering Information:
Available in five colors for easy identification of specimens: pink, blue, green, yellow, and orange. Multi-packs contain one of each color.
Specifications
Color Yellow
Packaging Type 2/Cs.
Quantity 4 oz. per bottle