Learn More
Epredia™ Mounting Medium
Catalog No. 22050102
Change view
Click to view available options
Volume (English):
4 oz.
1 pt.
Quantity:
2/Cs.
6/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Quantity
22050102 1 pt. 2/Cs.
22110610 4 oz. 6/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22050102 Supplier Epredia™ Supplier No. 4111
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Store slides for prolonged periods of time without fading of stains with Epredia™ Mounting Medium.
  • Rapid drying
  • Toluene-based
  • Optically clear
Ordering Alerts: Packaged in two configurations for convenience: a 1-pt. regular bottle or a 4-oz. plastic squeeze bottle with a twist-open/twist-close cap.
Specifications
Volume (English) 1 pt.
Quantity 2/Cs.
Product Type Mounting Medium