Learn More
Epredia™ Richard-Allan Scientific™ Modified Wright Stain Pack
Achieve optimum performance on Hema-Tek™ Slide Stainers with the Epredia™ Richard-Allan Scientific™ Stain Pack.
$1090.87 - $1287.42
Specifications
Quantity 5 Pk.
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity  
Catalog Number Mfr. No. Type Price Quantity  
22-050-268
View Documents
Epredia™
SH3000
Modified Wright Stain Pack
Case of 5 Each for $1,090.87
Only null left
22-050-269
View Documents
Epredia™
SH3005
Case of 5 Each for $1,287.42
Only null left
Description
Description
This system consists of a modified polychrome methylene blue-eosin stain (similar to Wright or Giemsa) and based on the original stain proposed by Romanowsky*.
Specifications
Specifications
5 Pk.
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'