Learn More
Epredia™ Richard-Allan Scientific™ Modified Grocott's Methenamine Silver (Chromic Acid) Kit
Uniquely modified silver stain formulation.
$422.68 - $1810.63
Specifications
| Type | Stain Kit |
|---|---|
| For Use With (Application) | Histological and Cytological Staining |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
88-041
|
Epredia™
88041 |
500 mL |
Each for $422.68
|
|
|||||
|
23-900-665
|
Epredia™
87024 |
1 Kit |
Each for $1,810.63
|
|
|||||
Description
- Utilizes methenamine borax capsules to make a working silver solution
- Distinguishes fungi, basement membranes, and opportunistic organisms in tissue sections
- Chromic acid solution enhances detection of Pneumocystis carinii, histoplasmosis, and fungus organisms; eliminates non-specific background staining
- Entities and reticular fibers stain black against a green background
Specifications
| Stain Kit | |
| Histological and Cytological Staining |