Learn More
Epredia™ Richard-Allan Scientific™ Methyl Green-Pyronin Y
Catalog No. 361014345
Change view
Click to view available options
Quantity:
125mL
Catalog No. Quantity
361014345 125mL
1 options
Catalog No. 361014345 Supplier Epredia™ Supplier No. 87011
Only null left
Effectively identify plasma cells and RNA in tissue sections and cytological preparations with Epredia™ Richard-Allan Scientific™ Methyl Green-Pyronin Y Stain
  • DNA is stained green
  • RNA is stained red
  • Concentrated MGP is diluted prior to use
Specifications
Type Methyl Green-Pyronin Y
Quantity 125mL