Learn More
Epredia™ Richard-Allan Scientific™ Iron Stain, Nuclear Red Thiazin stain solution
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Richard-Allan Scientific™ Iron Stain.
$240.03 - $586.65
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|
22-110-644
|
Epredia™
87006 |
Iron Stain Kit |
N/A
|
|
|||||
|
22-110-650
|
Epredia™
88014 |
Hydrochloric Acid Solution (20%) |
N/A
|
|
|||||
Description
- Useful in study of iron deficiency anemia, hemochromatosis and hemosiderosis
- Iron stains blue, nuclei stains pink-red, and cytoplasm stains pale pink