Learn More
Epredia™ Dehydrant Alcohol
Catalog No. 361025353
Change view
Click to view available options
Quantity:
1 gal.
1.32 gal.
Product Type:
Alcohol
Formulation:
100% Dehydrant
70% Dehydrant
80% Dehydrant
95% Dehydrant
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type Formulation
361025353 1 gal. Alcohol 70% Dehydrant
22050106 1 gal. Alcohol 100% Dehydrant
6215 1.32 gal. Alcohol 100% Dehydrant
22050107 1 gal. Alcohol 95% Dehydrant
6315 1.32 gal. Alcohol 95% Dehydrant
6415 1.32 gal. Alcohol 80% Dehydrant
6515 1.32 gal. Alcohol 70% Dehydrant
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 361025353 Supplier Epredia™ Supplier No. 6501
Only null left
Dehydrant Alcohol products are specifically designed for tissue processing and staining of histological and cytological specimens. Available in 70%, 80%, 95% and 100% blends for complete dehydration.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Specifications
Product Type Alcohol
Quantity 1 gal.
Formulation 70% Dehydrant