Learn More
Epredia™ Cytoseal™ Mountant
Catalog No. p-4532396
Change view
Click to view available options
Quantity:
4 oz.
16 oz.
Product Line:
Cytoseal 280
Cytoseal 60
Cytoseal XYL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Line
23244257 16 oz. Cytoseal 60
23244256 4 oz. Cytoseal 60
23244258 4 oz. Cytoseal 280
22050262 4 oz. Cytoseal XYL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 23244257 Supplier Epredia™ Supplier No. 831016
Only null left
Prevent cracking, fading and discoloration of stained specimens with Cytoseal™ mounting media.
  • Cytoseal 60 mountant is a low-viscosity medium that dries quickly and allows for rapid, even spreading which virtually eliminates air bubbles
Ordering Information:
Packaged in an easy-to-use dispenser with a twist-open/twist-closed cap.
Specifications
Product Line Cytoseal 60
Product Type Mountant
Quantity 16 oz.