Learn More
Epredia™ Cal-Rite™ Decalcifying/Fixation Solution
Catalog No. 22046339
Change view
Click to view available options
Quantity:
1 gal.
Catalog No. Quantity
22046339 1 gal.
1 options
Catalog No. 22046339 Supplier Epredia™ Supplier No. 5501
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Streamline processes with Cal-Rite Decalcifying/Fixation Solution, which allows simultaneous decalcification and fixation.
  • Beneficial for conventional iron staining
  • Preserves nuclear chromatin staining, even after prolonged exposure
  • Eliminates poor hematoxylin and eosin staining caused by over-decalcification and/or exposure to harsh mineral acids.
Specifications
Quantity 1 gal.