Learn More
Epredia™ Formaldehyde Warning Labels For Specimen Containers
Catalog No. 22046311
Change view
Click to view available options
Closure Included:
Yes
Catalog No. Closure Included
22046311 Yes
1 options
Catalog No. 22046311 Supplier Epredia™ Supplier No. SH4091
Two sizes of formaldehyde warning labels to complete any specimen collection needs.
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
  • Easy to transport trays ideal for operating rooms and hospitals
  • Clear polystyrene containers have screw-top lids
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
  • Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Easy to transport trays
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
  • Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
  • Stack pack reduces excess packaging
  • Comply with all updated federal and state OSHA requirements
  • Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
Product Type Specimen Container Label
Closure Included Yes
Description Formalin Warning Labels, Large