Learn More
Epredia™ Dry Specimen Containers - Snap Lid
Dry specimen containers constructed of clear and opaque polystyrene, polypropylene or polyethylene with a variety of lid options.
Supplier: Epredia™ 5333
Description
Three unique packaging configurations allow for easy distribution and transport.
53 Series Containers
- Easy to transport trays ideal for operating rooms and hospitals
- Clear polystyrene containers have screw-top lids
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
56 Series Containers
- Patented Epredia™ Bio-Tite™ closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Easy to transport trays
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
59 Series Containers
- Patented Epredia Bio-Tite closure system is dual-threaded, making it virtually impossible to cross-thread and minimizing leakage
- Stack pack reduces excess packaging
- Comply with all updated federal and state OSHA requirements
- Labeled with highly visible, bright yellow labels with space for patient information for instant identification to minimize health risks
Specifications
| Dry Specimen Container | |
| Polyethylene | |
| 1 L | |
| Opaque |
| Yes | |
| Non-sterile | |
| Snap Cap | |
| 100/Cs. |