Learn More
Epredia™ Base Mold Release
Catalog No. 361014317
Change view
Click to view available options
Quantity:
4 oz.
Catalog No. Quantity
361014317 4 oz.
1 options
Catalog No. 361014317 Supplier Epredia™ Supplier No. 6600
Only null left
Coat base molds for easy separation of paraffin from mold with Epredia™ Base Mold Release.
  • Paraffin buildup on the molds is reduced
  • Environmentally safe product is ready-to-use
  • Non-aerosol pump spray bottle
Specifications
Product Type Base Mold Release
Quantity 4 oz.