missing translation for 'onlineSavingsMsg'
Learn More

Eosinophil Stain, Eosin Y-Methylene Blue, for Mucous Secretions, Ricca Chemical

$348.59 - $375.41

Chemical Identifiers

CAS 67-56-1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
34-701-6
View Documents
Ricca Chemical Company
347016
500 mL
Each for $348.59
Only null left
 
34-703-2
View Documents
Ricca Chemical Company
347032
1 L
Each for $375.41
Only null left
 
Description

Description

Specifications

Chemical Identifiers

67-56-1

Specifications

67-56-1,7220-79-3,17372-87-1
Amber Glass Bottle
7.5
Eosinophil
Laboratory
Dark Blue
500 mL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'