missing translation for 'onlineSavingsMsg'
Learn More

Synthetic Urine Solution, Ricca Chemical

Catalog No. 83611
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
83611 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 83611 Supplier Ricca Chemical Company Supplier No. 83611

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General-use Reagent

  • Clear, colorless liquid
  • Specific gravity approximately 1
  • CAS: 57-13-6 (Urea) [1 - 3%]; 10034-99-8 (Magnesium Sulfate Heptahydrate) [<0.1%]; 10035-04-8 (Calcium Chloride Dihydrate) [<0.1%]; 7647-14-5 (Sodium Chloride) [<1%]; 7732-18-5 (Deionized Water) [balance]

Specifications

Certifications/Compliance NIST SRM 186, 191
Color Clear
Format Poly Bottle
For Use With (Equipment) General Analytical Equipment
Type Synthetic Urine
Grade Reagent
Includes Magnesium Sulfate Heptahydrate <0.1, Calcium Chloride Dihydrate <0.1, 1 to 3 Urea, Sodium Chloride, Water, Deionized
Packaging Natural Poly Bottle
pH 7.9
Quantity 4 L
Form Liquid
Show More Show Less