missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Electrode Cleaning Solution 2, Ricca Chemical
Description
- Clear, colorless to yellow liquid
- pH approx. 7.2 (10% solution)
- CAS: 57-13-6 (Urea); 7732-18-5 (Water, Deionized)
Specifications
Specifications
| Capacity (English) | 16.9 oz. |
| Capacity (Metric) | 500 mL |
| For Use With (Equipment) | Electrode |
| Type | Cleaning Solution |
| Packaging | Natural Poly Bottle |
| Quantity | 500 mL |