missing translation for 'onlineSavingsMsg'
Learn More

Copper Sulfate, Copper Sulfate, 1.055 Specific Gravity at 25°C (77°F), Ricca Chemical

For Hemoglobin Estimation in Men

Supplier:  Ricca Chemical Company RI23405

 View more versions of this product

Catalog No. 23405

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • 1.055 Specific Gravity at 25C (77F)
  • CAS: 7758-99-8 (Copper Sulfate Pentahydrate); 7732-18-5 (Water, Deionized)

Screening of male blood donors

Specifications

Chemical Identifiers

Screening of male Blood donors
4
15°C to 30°C

Specifications

15°C to 30°C
Cubitainer
20 L
Screening of male Blood donors
4
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 Months

SDS
Documents

Documents

Promotions

Promotions