missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulfate, Copper Sulfate, 1.055 Specific Gravity at 25°C (77°F), Ricca Chemical
For Hemoglobin Estimation in Men
Supplier: Ricca Chemical Company 234016
Description
- 1.055 Specific Gravity at 25C (77F)
- CAS: 7758-99-8 (Copper Sulfate Pentahydrate); 7732-18-5 (Water, Deionized)
Screening of male blood donors
Chemical Identifiers
| Screening of male Blood donors | |
| 4 | |
| 15°C to 30°C |
Specifications
| 15°C to 30°C | |
| Amber Glass | |
| 500 mL |
| Screening of male Blood donors | |
| 4 |
Safety and Handling
ShelfLife : 24 Months